Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001448.2 | NP_001439.2 | 3 | 850 | ||
GenBank mRNA | AF030186.1 | - | 3 | 850 | ||
AF064826.1 | - | 3 | 755 | |||
AK304207.1 | - | 3 | 575 | |||
AK312605.1 | - | 3 | 1107 | |||
AY052833.1 | - | 3 | 774 | |||
AY358507.1 | - | 3 | 1136 | |||
BC017166.1 | - | 3 | 1144 | 3922774 |