Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dysf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077694.2 | NP_001071162.1 | 3 | 535 | ||
NM_021469.3 | NP_067444.2 | 3 | 353 | |||
GenBank mRNA | AF188290.2 | - | 3 | 221 | ||
AK131144.1 | - | 3 | 350 | |||
CN716462.1 | - | 3 | 353 | |||
EF041824.1 | - | 3 | 498 |