Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163290.1 | NP_001156762.1 | 8 | 1032 | ||
NM_023341.3 | NP_075830.2 | 8 | 1130 | |||
GenBank mRNA | AJ278735.1 | - | 8 | 1010 | ||
AK004177.1 | - | 1 | 15 | |||
AK004791.1 | - | 8 | 1072 | |||
AK014605.2 | - | 8 | 1072 | |||
AK184581.1 | - | 5 | 404 | |||
BC030937.1 | - | 8 | 1078 | 4922892 | ||
U31629.1 | - | 6 | 671 |