Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133871.2 | NP_598632.2 | 2 | 400 | ||
GenBank mRNA | AJ299405.1 | - | 1 | 290 | ||
AK079749.1 | - | 2 | 352 | |||
AK085407.1 | - | 2 | 389 | |||
AK085650.1 | - | 2 | 334 | |||
AK136553.1 | - | 2 | 352 | |||
AK151346.1 | - | 2 | 352 | |||
AK171676.1 | - | 2 | 333 | |||
BB645803.1 | - | 2 | 389 | |||
BC111882.1 | - | 1 | 290 | 40060245 | ||
BC112402.1 | - | 1 | 290 |