siRNA Details
Gene Symbol: Ddhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039106.3 | NP_001034195.2 | 4 | 1263 | ||
NM_001042719.2 | NP_001036184.1 | 5 | 1365 | |||
NM_001284399.1 | NP_001271328.1 | 6 | 1337 | |||
NM_176845.5 | NP_789815.3 | 4 | 1263 | |||
GenBank mRNA | AK014975.1 | - | 2 | 113 | ||
AK048690.1 | - | 5 | 823 | |||
AK129425.1 | - | 2 | 674 | |||
AK155294.1 | - | 6 | 1294 | |||
AK170271.1 | - | 5 | 977 | |||
BC043475.1 | - | 3 | 517 | 5365658 | ||
CF532335.1 | - | 3 | 185 | |||
DQ272481.1 | - | 4 | 1262 | |||
DQ272482.1 | - | 4 | 1266 |

More Information
Other Species Targeted

Panther Classification:
