Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286809.1 | NP_001273738.1 | 3 | 478 | ||
NM_009792.3 | NP_033922.1 | 4 | 511 | |||
NM_177407.4 | NP_803126.1 | 15 | 1199 | |||
GenBank mRNA | AK037463.1 | - | 1 | 718 | ||
AK083245.1 | - | 15 | 1200 | |||
AK122410.2 | - | 15 | 1195 | |||
BB659027.1 | - | 3 | 434 | |||
BC031745.1 | - | 15 | 1180 | 4506005 | ||
X14836.1 | - | 15 | 1183 | |||
X87142.1 | - | 4 | 511 |