Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040398.2 | NP_001035488.2 | 9, 10 | 3584 | ||
GenBank mRNA | BC035291.1 | - | 1, 2 | 86 | ||
BC038367.1 | - | 3, 4 | 662 | |||
BC040775.1 | - | 1, 2 | 86 | |||
BC041681.1 | - | 1, 2 | 105 |