Search
Search

Ptbp3| Gene Symbol: Ptbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144904.2 | NP_659153.2 | 14 | 1723 | ||
| NM_178164.3 | NP_835458.1 | 13 | 1698 | |||
| GenBank mRNA | AK033732.1 | - | 13 | 1623 | ||
| AK042920.1 | - | 11 | 1630 | |||
| AK077791.1 | - | 1 | 69 | |||
| AK088560.1 | - | 13 | 1630 | |||
| BC006638.1 | - | 14 | 1633 | 3969488 | ||
| BC057641.1 | - | 13 | 1603 | 4936557 | ||