Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhlhe23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080641.5 | NP_542372.2 | 1 | 579 | ||
GenBank mRNA | AF454760.1 | - | 1 | 428 | ||
AK044215.1 | - | 1 | 580 | |||
AK051155.1 | - | 1 | 580 | |||
BC119208.1 | - | 1 | 489 | 8733957 | ||
BC119210.1 | - | 1 | 488 | 8733959 |