siRNA Details
Gene Symbol: Pou3f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011141.2 | NP_035271.1 | 1 | 1128 | ||
GenBank mRNA | X54628.1 | - | 1 | 1121 | ||
X56959.1 | - | 1 | 1102 | |||
X57482.1 | - | 1 | 1086 |

More Information
Other Species Targeted

