Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016844.2 | NP_058540.1 | 3 | 128 | ||
GenBank mRNA | AK002919.1 | - | 3 | 126 | ||
AK089360.1 | - | 1 | 1180 | |||
AV497904.1 | - | 3 | 107 | |||
BC010987.1 | - | 3 | 123 | 4218716 | ||
BC090982.1 | - | 3 | 125 | 30316901 | ||
BC125644.1 | - | 3 | 192 | 40130159 | ||
BC125646.1 | - | 3 | 192 | 40130161 | ||
BY502260.1 | - | 3 | 131 | |||
U11248.1 | - | 3 | 109 |