Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atoh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016864.1 | NP_058560.1 | 1 | 233 | ||
GenBank mRNA | AF071223.1 | - | 1 | 233 | ||
AK005214.1 | - | 1 | 26 | |||
BC092234.1 | - | 1 | 586 | 30940279 |