Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024414.2 | NP_077725.1 | 2 | 326 | ||
GenBank mRNA | BC132042.1 | - | 2 | 121 | 40130319 | |
BC132068.1 | - | 2 | 121 | 40130345 | ||
D29743.1 | - | 2 | 153 |