siRNA Details
Gene Symbol: Sox21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177753.3 | NP_808421.1 | 1 | 1616 | ||
GenBank mRNA | AY069926.1 | - | 1 | 435 | ||
AY142959.1 | - | 1 | 488 | |||
BC063054.1 | - | 1 | 676 | |||
BC069176.1 | - | 2 | 917 | |||
BC147556.1 | - | 1 | 383 | 9056548 | ||
BC147565.1 | - | 1 | 383 | 9056555 |
