Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146966.2 | NP_667177.2 | 1 | 439 | ||
GenBank mRNA | BC125540.1 | - | 1 | 433 | 40130055 | |
BC132105.1 | - | 1 | 433 | 40130382 |