siRNA Details
Gene Symbol: Alx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007442.3 | NP_031468.1 | 1 | 359 | ||
GenBank mRNA | AB221573.1 | - | 1 | 138 | ||
AF001465.1 | - | 2 | 329 | |||
BC049786.1 | - | 1 | 427 |

More Information
Other Species Targeted

Panther Classification:
