Search
Search

BTC| Gene Symbol: BTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316963.1 | NP_001303892.1 | 2, 3 | 519 | ||
| NM_001729.3 | NP_001720.1 | 2, 3 | 519 | |||
| XM_011532211.1 | XP_011530513.1 | 2, 3 | 500 | |||
| GenBank mRNA | BC011618.2 | - | 2, 3 | 310 | 3952444 | |
| BI552049.1 | - | 2, 3 | 335 | |||
| KU177946.1 | - | 2, 3 | 158 | |||
| KU177947.1 | - | 2, 3 | 158 | |||
| S55606.1 | - | 2, 3 | 452 | |||