Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pura | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008989.3 | NP_033015.1 | 2 | 985 | ||
GenBank mRNA | AF017631.1 | - | 1 | 510 | ||
U02098.1 | - | 2 | 985 |