Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CIB1 SIRNA Assays ›
          Gene Symbol:
          Cib1
          Gene Name:
          calcium and integrin binding 1 (calmyrin)
          Gene Aliases:
          Cibkip, Kip, Prkdcip
          Chromosome Location:
          Chr.7: 80227156 - 80232805 on Build GRCm38
          UniGene ID:
          Mm.30217
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 102842

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cib1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291275.1 NP_001278204.1 3 410
          NM_011870.5 NP_036000.1 3 410
          GenBank mRNA AB006463.1 - 3 191
          AF173010.1 - 3 189
          AK010345.1 - 3 207
          AK027915.1 - 3 222
          AK152805.1 - 3 242
          AK165472.1 - 3 195
          BC003714.1 - 3 196 3599986
          BC054385.1 - 3 215 6476137
          CF104297.1 - 3 418

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          positive regulation of cell-matrix adhesion
          response to ischemia
          negative regulation of protein kinase activity
          apoptotic process
          cellular response to DNA damage stimulus
          negative regulation of microtubule depolymerization
          cell cycle
          endomitotic cell cycle
          cell adhesion
          spermatogenesis
          spermatid development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          negative regulation of neuron projection development
          cell differentiation
          platelet formation
          positive regulation of cell growth
          positive regulation of cell migration
          cytoplasmic microtubule organization
          positive regulation of cell adhesion mediated by integrin
          thrombopoietin-mediated signaling pathway
          regulation of cell proliferation
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          negative regulation of megakaryocyte differentiation
          positive regulation of NF-kappaB transcription factor activity
          cell division
          regulation of cell division
          negative regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of calcineurin-NFAT signaling cascade
          cellular response to tumor necrosis factor
          cellular response to growth factor stimulus
          negative regulation of protein serine/threonine kinase activity
          positive regulation of protein serine/threonine kinase activity
          positive regulation of establishment of protein localization to plasma membrane
          positive regulation of cell migration involved in sprouting angiogenesis
          positive regulation of protein targeting to membrane
          positive regulation of substrate adhesion-dependent cell spreading
          cellular response to nerve growth factor stimulus
          positive regulation of male germ cell proliferation
          calcium ion binding
          protein binding
          calcium-dependent protein kinase inhibitor activity
          Ras GTPase binding
          protein serine/threonine kinase inhibitor activity
          protein anchor
          ion channel binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0