Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256030.1 | NP_001242959.1 | 3 | 618 | ||
NM_001778.3 | NP_001769.2 | 3 | 618 | |||
XM_011510171.1 | XP_011508473.1 | 4 | 686 | |||
XM_017002867.1 | XP_016858356.1 | 2 | 302 | |||
GenBank mRNA | AK225636.1 | - | 3 | 591 | ||
AK310624.1 | - | 3 | 592 | |||
AK310744.1 | - | 3 | 598 | |||
BC016182.1 | - | 3 | 562 | 4850933 | ||
BT019813.1 | - | 3 | 530 | |||
CR457012.1 | - | 3 | 530 | |||
M37766.1 | - | 3 | 550 | |||
M59904.1 | - | 3 | 547 | |||
X06341.1 | - | 3 | 566 |