Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT7 SIRNA Assays ›
Gene Symbol:
SEPT7
Gene Name:
septin 7
Gene Aliases:
CDC10, CDC3, NBLA02942, SEPT7A
Chromosome Location:
Chr.7: 35800986 - 35915763 on Build GRCh38
UniGene ID:
Hs.191346
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
10323
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001011553.3
NP_001011553.2
2
371
NM_001788.5
NP_001779.3
3
374
XM_006715806.3
XP_006715869.1
3
322
XM_011515656.2
XP_011513958.1
3
322
XM_011515661.2
XP_011513963.1
3
322
XM_017012855.1
XP_016868344.1
3
306
XM_017012856.1
XP_016868345.1
3
314
XM_017012857.1
XP_016868346.1
3
311
XM_017012858.1
XP_016868347.1
3
142
XM_017012859.1
XP_016868348.1
2
139
XM_017012860.1
XP_016868349.1
3
136
XM_017012861.1
XP_016868350.1
3
133
XM_017012862.1
XP_016868351.1
2
137
XM_017012863.1
XP_016868352.1
3
680
GenBank mRNA
AB073389.1
-
3
141
AB209677.1
-
3
109
AK223225.1
-
3
141
AK223241.1
-
3
141
AK290545.1
-
2
212
AK298453.1
-
3
208
BC025987.2
-
2
138
4823810
BC067264.1
-
2
117
6503495
BC093640.1
-
3
140
7939485
BC093642.1
-
3
140
7939487
BC107718.1
-
2
319
BI669760.1
-
3
352
BP280069.1
-
3
345
BQ229116.1
-
2
302
BQ232357.1
-
3
116
BX571755.1
-
2
139
DC414394.1
-
2
374
DQ232879.1
-
2
122
KF305319.1
-
3
112
S72008.1
-
3
141
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cytokinesis
mitotic nuclear division
regulation of embryonic cell shape
protein heterooligomerization
cilium morphogenesis
cell-cell adhesion
positive regulation of nonmotile primary cilium assembly
structural molecule activity
protein binding
GTP binding
identical protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-zbtnl:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0