siRNA Details
Gene Symbol: Rbbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011247.2 | NP_035377.2 | 1 | 513 | ||
NM_175023.3 | NP_778188.1 | 1 | 513 | |||
GenBank mRNA | AK045635.1 | - | 1 | 258 | ||
AK081261.1 | - | 1 | 377 | |||
BC025874.1 | - | 1 | 150 | |||
BE994197.1 | - | 1 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
