Search Thermo Fisher Scientific
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020247.4 | NP_064632.2 | 15 | 2388 | ||
XM_005273201.1 | XP_005273258.1 | 15 | 2284 | |||
XM_011544238.1 | XP_011542540.1 | 16 | 2480 | |||
XM_011544239.2 | XP_011542541.1 | 17 | 2503 | |||
XM_011544240.2 | XP_011542542.1 | 15 | 2297 | |||
XM_011544241.2 | XP_011542543.1 | 15 | 2259 | |||
XM_017001852.1 | XP_016857341.1 | 15 | 2478 | |||
GenBank mRNA | AB073905.1 | - | 15 | 2311 | ||
AF218003.1 | - | 7 | 1403 | |||
AJ278126.1 | - | 11 | 1606 | |||
AK074693.1 | - | 15 | 2331 | |||
AK090494.1 | - | 15 | 2125 | |||
AK092784.1 | - | 15 | 2111 | |||
AK126200.1 | - | 8 | 3076 | |||
AK297735.1 | - | 12 | 1667 | |||
AK301012.1 | - | 17 | 2309 | |||
BC005171.2 | - | 15 | 2337 | 3604948 | ||
BX648860.1 | - | 15 | 2209 |