Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHEK2 SIRNA Assays ›
          Gene Symbol:
          CHEK2
          Gene Name:
          checkpoint kinase 2
          Gene Aliases:
          CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1
          Chromosome Location:
          Chr.22: 28687743 - 28741866 on Build GRCh38
          UniGene ID:
          Hs.291363
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103344

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHEK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005735.1 NP_001005735.1 4 559
          NM_001257387.1 NP_001244316.1 3 430
          NM_007194.3 NP_009125.1 3 430
          NM_145862.2 NP_665861.1 3 430
          XM_011529839.2 XP_011528141.1 4 581
          XM_011529840.2 XP_011528142.1 4 583
          XM_011529841.1 XP_011528143.1 4 563
          XM_011529842.2 XP_011528144.1 3 451
          XM_011529843.1 XP_011528145.1 3 430
          XM_011529844.2 XP_011528146.1 4 591
          XM_011529845.2 XP_011528147.1 2 137
          XM_017028560.1 XP_016884049.1 4 545
          GenBank mRNA AB040105.1 - 2 358
          AF086904.1 - 2 358
          AF096279.1 - 2 358
          AF174135.1 - 2 358
          AF217975.1 - 3 1029
          AJ131197.1 - 2 358
          AJ783839.1 - 2 358
          AK289360.1 - 3 409
          AK290754.1 - 3 432
          AY551296.1 - 2 358
          AY551297.1 - 3 487
          AY551298.1 - 2 358
          AY551299.1 - 2 358
          AY551300.1 - 2 358
          AY551301.1 - 2 358
          AY551302.1 - 2 358
          AY551305.1 - 2 358
          BC004207.2 - 3 385 3532866
          BM838597.1 - 3 430
          CR456418.1 - 4 558

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          G2/M transition of mitotic cell cycle
          replicative cell aging
          positive regulation of protein phosphorylation
          double-strand break repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          cellular response to DNA damage stimulus
          DNA damage induced protein phosphorylation
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to gamma radiation
          peptidyl-serine phosphorylation
          cellular response to drug
          regulation of protein catabolic process
          signal transduction in response to DNA damage
          cellular protein catabolic process
          positive regulation of transcription, DNA-templated
          protein autophosphorylation
          protein stabilization
          cell division
          negative regulation of cell cycle arrest
          signal transduction involved in intra-S DNA damage checkpoint
          mitotic spindle assembly
          replicative senescence
          regulation of signal transduction by p53 class mediator
          response to doxorubicin
          cellular response to bisphenol A
          negative regulation of DNA damage checkpoint
          positive regulation of anoikis
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          protein kinase binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0