Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007371.3 | NP_031397.1 | 4 | 625 | ||
XM_006717291.2 | XP_006717354.1 | 4 | 757 | |||
XM_011519052.2 | XP_011517354.1 | 4 | 976 | |||
XM_017015165.1 | XP_016870654.1 | 3 | 550 | |||
GenBank mRNA | AK299519.1 | - | 4, 5 | 757 | ||
AY513270.1 | - | 3 | 455 | |||
BC032124.2 | - | 4 | 735 | 4856840 | ||
BI545861.1 | - | 4 | 635 | |||
D26362.1 | - | 4 | 576 | |||
KR633047.1 | - | 3 | 437 |