Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EPHA7 SIRNA Assays ›
          Gene Symbol:
          EPHA7
          Gene Name:
          EPH receptor A7
          Gene Aliases:
          EHK-3, EHK3, EK11, HEK11
          Chromosome Location:
          Chr.6: 93240020 - 93419687 on Build GRCh38
          UniGene ID:
          Hs.73962
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103405

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EPHA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001288629.1 NP_001275558.1 5 1437
          NM_004440.3 NP_004431.1 5 1437
          XM_005248669.2 XP_005248726.1 5 1494
          XM_005248671.2 XP_005248728.1 5 1494
          XM_017010365.1 XP_016865854.1 5 1494
          XM_017010366.1 XP_016865855.1 5 1494
          GenBank mRNA AB209269.1 - 5 1424
          AK313529.1 - 5 1409
          BC126125.1 - 5 1311 8991841
          BC126151.1 - 5 1311 8991867
          BC143857.1 - 5 1311 9052373
          BC143858.1 - 5 1311 9052374
          L36642.1 - 5 1381

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_134331, XM_006237965, XM_006237962, XM_006237963, XM_008763587, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001122889, NM_010141, NM_001290434, BC026153, X79082, and others.

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          apoptotic process
          brain development
          phosphorylation
          peptidyl-tyrosine phosphorylation
          regulation of cell-cell adhesion
          retinal ganglion cell axon guidance
          regulation of protein autophosphorylation
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of neuron apoptotic process
          ephrin receptor signaling pathway
          negative regulation of collateral sprouting
          branching morphogenesis of a nerve
          regulation of peptidyl-tyrosine phosphorylation
          negative chemotaxis
          negative regulation of synapse assembly
          regulation of ERK1 and ERK2 cascade
          nephric duct morphogenesis
          protein tyrosine kinase activity
          GPI-linked ephrin receptor activity
          protein binding
          ATP binding
          axon guidance receptor activity
          chemorepellent activity
          ephrin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0