Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145443.2 | NP_001138915.1 | 10 | 1118 | ||
NM_001282630.2 | NP_001269559.1 | 10 | 1231 | |||
NM_001314063.1 | NP_001300992.1 | 10 | 1442 | |||
NM_001323016.1 | NP_001309945.1 | 10 | 1118 | |||
NM_001323017.1 | NP_001309946.1 | 10 | 1492 | |||
NM_004566.3 | NP_004557.1 | 10 | 1442 | |||
XM_005252464.1 | XP_005252521.1 | 10 | 1442 | |||
XM_017016325.1 | XP_016871814.1 | 10 | 1442 | |||
XM_017016326.1 | XP_016871815.1 | 9 | 1385 | |||
XM_017016327.1 | XP_016871816.1 | 10 | 1194 | |||
XM_017016329.1 | XP_016871818.1 | 9 | 1385 | |||
GenBank mRNA | AF056320.1 | - | 11 | 1102 | ||
AF109735.1 | - | 10 | 1172 | |||
AK291263.1 | - | 10 | 1387 | |||
AK303039.1 | - | 10 | 1051 | |||
AK304450.1 | - | 10 | 1163 | |||
BC037258.1 | - | 10 | 1043 | |||
BC040482.1 | - | 10 | 1387 | 5271907 | ||
D49817.1 | - | 10 | 1076 | |||
L77662.1 | - | 10 | 1396 |