Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP2B4 SIRNA Assays ›
          Gene Symbol:
          ATP2B4
          Gene Name:
          ATPase plasma membrane Ca2+ transporting 4
          Gene Aliases:
          ATP2B2, MXRA1, PMCA4, PMCA4b, PMCA4x
          Chromosome Location:
          Chr.1: 203626787 - 203744081 on Build GRCh38
          UniGene ID:
          Hs.343522
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 10353

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATP2B4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001396.2 NP_001001396.1 3 1139
          NM_001684.4 NP_001675.3 3 1139
          GenBank mRNA AK292458.1 - 2 478
          BC140774.1 - 3 811 9021642
          BC150293.1 - 2 496 8860318
          BX537444.1 - 3 1366
          CR749393.1 - 3 1069
          CR936708.1 - 3 1093
          M25874.1 - 2 639
          M83363.1 - 2 639

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          neural retina development
          regulation of transcription from RNA polymerase II promoter
          cellular calcium ion homeostasis
          spermatogenesis
          negative regulation of nitric oxide mediated signal transduction
          hippocampus development
          sperm motility
          positive regulation of peptidyl-serine phosphorylation
          ion transmembrane transport
          negative regulation of nitric oxide biosynthetic process
          negative regulation of nitric-oxide synthase activity
          response to hydrostatic pressure
          calcium ion transmembrane transport
          negative regulation of calcineurin-NFAT signaling cascade
          cellular response to epinephrine stimulus
          calcium ion transmembrane import into cytosol
          calcium ion import across plasma membrane
          negative regulation of the force of heart contraction
          negative regulation of arginine catabolic process
          negative regulation of adrenergic receptor signaling pathway involved in heart process
          calcium ion export
          negative regulation of peptidyl-cysteine S-nitrosylation
          regulation of sodium ion transmembrane transport
          regulation of cell cycle G1/S phase transition
          negative regulation of cardiac muscle hypertrophy in response to stress
          negative regulation of citrulline biosynthetic process
          regulation of cardiac conduction
          positive regulation of cAMP-dependent protein kinase activity
          calcium-transporting ATPase activity
          protein binding
          calmodulin binding
          ATP binding
          sodium channel regulator activity
          PDZ domain binding
          protein phosphatase 2B binding
          nitric-oxide synthase inhibitor activity
          metal ion binding
          nitric-oxide synthase binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0