Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MET SIRNA Assays ›
          Gene Symbol:
          MET
          Gene Name:
          MET proto-oncogene, receptor tyrosine kinase
          Gene Aliases:
          AUTS9, DFNB97, HGFR, RCCP2, c-Met
          Chromosome Location:
          Chr.7: 116672359 - 116798386 on Build GRCh38
          UniGene ID:
          Hs.132966
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103545

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MET
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000245.3 NP_000236.2 2 450
          NM_001127500.2 NP_001120972.1 2 450
          NM_001324401.1 NP_001311330.1 2 450
          XM_011516223.1 XP_011514525.1 3 479
          GenBank mRNA BC130420.1 - 2 413 40146451
          DB223186.1 - 3 511
          DQ656040.2 - 3 484
          EU176015.1 - 2 442
          EU826567.1 - 1 248
          EU826568.1 - 1 248
          EU826569.1 - 1 248
          EU826570.1 - 1 248
          EU826571.1 - 1 248
          EU826572.1 - 1 248
          EU826573.1 - 1 248
          EU826574.1 - 1 248
          EU826575.1 - 1 248
          EU826576.1 - 1 248
          EU826577.1 - 1 248
          EU826578.1 - 1 248
          EU826579.1 - 1 248
          EU826580.1 - 1 248
          EU826581.1 - 1 248
          J02958.1 - 2 442
          X54559.1 - 2 442

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          endothelial cell morphogenesis
          signal transduction
          cell surface receptor signaling pathway
          cell proliferation
          negative regulation of autophagy
          peptidyl-tyrosine phosphorylation
          positive regulation of transcription from RNA polymerase II promoter
          hepatocyte growth factor receptor signaling pathway
          branching morphogenesis of an epithelial tube
          positive chemotaxis
          semaphorin-plexin signaling pathway
          negative regulation of hydrogen peroxide-mediated programmed cell death
          positive regulation of endothelial cell chemotaxis
          protein tyrosine kinase activity
          hepatocyte growth factor-activated receptor activity
          protein binding
          ATP binding
          protein phosphatase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0