Search
Search

ERBB2| Gene Symbol: ERBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005862.2 | NP_001005862.1 | 21 | 2660 | ||
| NM_001289936.1 | NP_001276865.1 | 22 | 2711 | |||
| NM_001289937.1 | NP_001276866.1 | 18 | 2435 | |||
| NM_004448.3 | NP_004439.2 | 18 | 2435 | |||
| NR_110535.1 | 18 | 2498 | ||||
| GenBank mRNA | AK131568.1 | - | 14 | 3308 | ||
| AK295195.1 | - | 22 | 2711 | |||
| AK300321.1 | - | 12 | 1609 | |||
| AK313683.1 | - | 18 | 2389 | |||
| BC080193.1 | - | 18 | 2396 | |||
| BC167147.1 | - | 18 | 2177 | 100013444 | ||
| HM437238.1 | - | 1 | 24 | |||
| KJ534831.1 | - | 18 | 2174 | |||
| KJ534964.1 | - | 17 | 2126 | |||
| M11730.1 | - | 18 | 2324 | |||
| X03363.1 | - | 18 | 2348 | |||