siRNA Details
Gene Symbol: OPN1LW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020061.5 | NP_064445.2 | 5 | 821 | ||
GenBank mRNA | AW950066.1 | - | 2 | 185 | ||
BM688032.1 | - | 2 | 49 | |||
BQ637240.1 | - | 4 | 495 | |||
CR749814.1 | - | 5 | 779 | |||
EL949562.1 | - | 3 | 330 |
Gene Symbol: OPN1MW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000513.2 | NP_000504.1 | 5 | 843 | ||
GenBank mRNA | BC140938.1 | - | 5 | 843 | 9056750 | |
BC143790.1 | - | 5 | 824 | 9052304 | ||
BC171754.1 | - | 5 | 824 | 9053255 | ||
BC171755.1 | - | 5 | 824 | 9053256 |
Gene Symbol: OPN1MW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048181.2 | NP_001041646.1 | 5 | 843 |
Gene Symbol: OPN1MW3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003960089.4 | XP_003960138.1 | 5 | 761 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Panther Classification:
