Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146068.1 | NP_001139540.1 | 3 | 406 | ||
NM_001748.4 | NP_001739.2 | 3 | 628 | |||
XM_011544286.2 | XP_011542588.1 | 3 | 805 | |||
GenBank mRNA | AB209855.1 | - | 3 | 493 | ||
AF261089.1 | - | 3 | 531 | |||
AK297800.1 | - | 3 | 406 | |||
AK315179.1 | - | 3 | 530 | |||
AK316211.1 | - | 3 | 449 | |||
BC007686.2 | - | 3 | 628 | |||
BC011828.2 | - | 3 | 628 | |||
BC021303.2 | - | 3 | 522 | 5090865 | ||
M23254.1 | - | 3 | 534 |