Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ECE1 SIRNA Assays ›
          Gene Symbol:
          ECE1
          Gene Name:
          endothelin converting enzyme 1
          Gene Aliases:
          ECE
          Chromosome Location:
          Chr.1: 21217247 - 21345541 on Build GRCh38
          UniGene ID:
          Hs.195080
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103998

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ECE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113347.1 NP_001106818.1 10 1689
          NM_001113348.1 NP_001106819.1 12 1584
          NM_001113349.1 NP_001106820.1 11 1535
          NM_001397.2 NP_001388.1 12 1544
          XM_006710398.2 XP_006710461.1 12 1689
          XM_011540872.2 XP_011539174.1 12 2296
          XM_011540873.2 XP_011539175.1 12 1614
          XM_017000511.1 XP_016856000.1 12 1436
          GenBank mRNA AB031742.1 - 11 1429
          AK290656.1 - 12 1516
          AK296486.1 - 12 1519
          AK304167.1 - 11 1477
          AK309677.1 - 12 1518
          BC117256.1 - 12 1544 40125807
          BC126257.1 - 12 1544 8991973
          BX648351.1 - 10 1548
          CN298523.1 - 4 441
          CN298525.1 - 3 226
          D43698.1 - 10 1494
          D49471.1 - 12 1469
          Z35307.1 - 12 1455

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of receptor recycling
          regulation of systemic arterial blood pressure by endothelin
          apoptotic process
          heart development
          positive regulation of cardiac muscle hypertrophy
          substance P catabolic process
          bradykinin catabolic process
          calcitonin catabolic process
          protein processing
          peptide hormone processing
          regulation of vasoconstriction
          positive regulation of cAMP biosynthetic process
          endothelin maturation
          response to muscle stretch
          regulation of vasodilation
          hormone catabolic process
          response to drug
          embryonic digit morphogenesis
          ear development
          positive regulation of G-protein coupled receptor protein signaling pathway
          response to cadmium ion
          pharyngeal system development
          regulation of ERK1 and ERK2 cascade
          endopeptidase activity
          metalloendopeptidase activity
          protein binding
          peptide hormone binding
          protein homodimerization activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0