Search
Search

ADAM12| Gene Symbol: ADAM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288973.1 | NP_001275902.1 | 12, 13 | 1744 | ||
| NM_001288974.1 | NP_001275903.1 | 12, 13 | 1744 | |||
| NM_001288975.1 | NP_001275904.1 | 12, 13 | 1744 | |||
| NM_003474.5 | NP_003465.3 | 12, 13 | 1753 | |||
| NM_021641.4 | NP_067673.2 | 12, 13 | 1753 | |||
| XM_017016705.1 | XP_016872194.1 | 11, 12 | 1656 | |||
| XM_017016706.1 | XP_016872195.1 | 2, 3 | 795 | |||
| GenBank mRNA | AF023476.2 | - | 12, 13 | 1646 | ||
| AF023477.2 | - | 12, 13 | 1646 | |||
| AK127856.1 | - | 12, 13 | 1649 | |||
| AK291629.1 | - | 12, 13 | 1563 | |||
| AY358878.1 | - | 12, 13 | 1641 | |||
| BC060804.1 | - | 12, 13 | 1683 | 30348514 | ||
| KF444157.1 | - | 12, 13 | 1744 | |||