Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD63 SIRNA Assays ›
          Gene Symbol:
          CD63
          Gene Name:
          CD63 molecule
          Gene Aliases:
          LAMP-3, ME491, MLA1, OMA81H, TSPAN30
          Chromosome Location:
          Chr.12: 55725443 - 55729673 on Build GRCh38
          UniGene ID:
          Hs.445570
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 10412

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD63
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257389.1 NP_001244318.1 4, 5 741
          NM_001257390.1 NP_001244319.1 4, 5 484
          NM_001257391.1 NP_001244320.1 3, 4 479
          NM_001257392.1 NP_001244321.1 3, 4 280
          NM_001257400.1 NP_001244329.1 3, 4 309
          NM_001257401.1 NP_001244330.1 3, 4 449
          NM_001267698.1 NP_001254627.1 4, 5 531
          NM_001780.5 NP_001771.1 4, 5 535
          GenBank mRNA AF508304.1 - 3, 4 277
          AK311893.1 - 4, 5 406
          AL582859.3 - 3, 4 354
          BC002349.2 - 4, 5 423 2819554
          BC013017.2 - 4, 5 379 3010594
          BE407235.1 - 3, 4 478
          BG478842.1 - 4, 5 485
          BQ434569.1 - 450
          BT007073.1 - 3, 4 338
          BT020137.1 - 3, 4 338
          BT020138.1 - 3, 4 338
          BU167765.1 - 4, 5 542
          BU190521.1 - 3, 4 474
          CN296813.1 - 4, 5 498
          CN484590.1 - 3, 4 349
          CR542096.1 - 3, 4 338
          EB386351.1 - 4, 5 470
          KF998086.1 - 3, 4 338
          M58485.1 - 4, 5 393
          M59907.1 - 4, 5 388
          S93788.1 - 3, 4 362
          X07982.1 - 4, 5 365

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor internalization
          platelet degranulation
          cell-matrix adhesion
          negative regulation of epithelial cell migration
          protein transport
          cell migration
          epithelial cell differentiation
          cellular protein localization
          endosome to melanosome transport
          positive regulation of cell adhesion
          pigment granule maturation
          regulation of vascular endothelial growth factor signaling pathway
          regulation of rubidium ion transport
          positive regulation of integrin-mediated signaling pathway
          protein binding
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0