Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543544.1 | XP_011541846.1 | 13 | 2217 | ||
GenBank mRNA | AB109031.1 | - | 14 | 2253 | ||
AB163917.1 | - | 14 | 2309 | |||
AF191545.1 | - | 13 | 2225 | |||
AK074387.1 | - | 7 | 829 | |||
AK312864.1 | - | 14 | 2266 | |||
AY028805.1 | - | 13 | 2239 | |||
BC065240.1 | - | 13 | 2112 | 6166289 |