Search
Search

KCNA6| Gene Symbol: KCNA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002235.3 | NP_002226.1 | 1 | 2598 | ||
| XM_005253686.3 | XP_005253743.1 | 1 | 4897 | |||
| XM_011520955.2 | XP_011519257.1 | 1 | 4897 | |||
| XM_017019270.1 | XP_016874759.1 | 1 | 4897 | |||
| XM_017019271.1 | XP_016874760.1 | 1 | 4897 | |||
| XM_017019272.1 | XP_016874761.1 | 1 | 4897 | |||
| GenBank mRNA | X17622.1 | - | 1 | 2594 | ||