Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN1 SIRNA Assays ›
          Gene Symbol:
          PTPN1
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 1
          Gene Aliases:
          PTP1B
          Chromosome Location:
          Chr.20: 50510321 - 50584762 on Build GRCh38
          UniGene ID:
          Hs.417549
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104533
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278618.1 NP_001265547.1 6 953
          NM_002827.3 NP_002818.1 7 1044
          GenBank mRNA AK290638.1 - 7 1011
          AK299830.1 - 6 891
          AK316563.1 - 7 986
          BC015660.2 - 7 973 4844022
          BC018164.1 - 7 954 4338750
          BT006752.1 - 7 837
          M31724.1 - 7 927
          M33689.1 - 7 909

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00182260_m1

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          activation of signaling protein activity involved in unfolded protein response
          activation of JUN kinase activity
          insulin receptor signaling pathway
          regulation of signal transduction
          negative regulation of signal transduction
          regulation of endocytosis
          negative regulation of vascular endothelial growth factor receptor signaling pathway
          endoplasmic reticulum unfolded protein response
          actin cytoskeleton reorganization
          regulation of intracellular protein transport
          peptidyl-tyrosine dephosphorylation
          platelet-derived growth factor receptor-beta signaling pathway
          IRE1-mediated unfolded protein response
          negative regulation of MAP kinase activity
          negative regulation of insulin receptor signaling pathway
          regulation of type I interferon-mediated signaling pathway
          JAK-STAT cascade involved in growth hormone signaling pathway
          positive regulation of protein tyrosine kinase activity
          negative regulation of ERK1 and ERK2 cascade
          cell-cell adhesion
          regulation of hepatocyte growth factor receptor signaling pathway
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          positive regulation of IRE1-mediated unfolded protein response
          negative regulation of PERK-mediated unfolded protein response
          peptidyl-tyrosine dephosphorylation involved in inactivation of protein kinase activity
          positive regulation of receptor catabolic process
          protein tyrosine phosphatase activity
          insulin receptor binding
          protein binding
          zinc ion binding
          enzyme binding
          protein kinase binding
          receptor tyrosine kinase binding
          poly(A) RNA binding
          ephrin receptor binding
          protein phosphatase 2A binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0