Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270422.1 | NP_001257351.1 | 6 | 1797 | ||
NM_001291622.1 | NP_001278551.1 | 6 | 1751 | |||
NM_001291623.1 | NP_001278552.1 | 3 | 1544 | |||
NM_001291624.1 | NP_001278553.1 | 4 | 1672 | |||
NM_001291625.1 | NP_001278554.1 | 5 | 1753 | |||
NM_018658.2 | NP_061128.2 | 5 | 1669 | |||
NM_170741.2 | NP_733937.2 | 4 | 1588 | |||
NM_170742.2 | NP_733938.2 | 4 | 1625 | |||
XM_005257337.4 | XP_005257394.1 | 3 | 1471 | |||
XM_006721885.3 | XP_006721948.1 | 7 | 1877 | |||
XM_006721886.3 | XP_006721949.1 | 6 | 1749 | |||
XM_006721887.3 | XP_006721950.1 | 6 | 1742 | |||
XM_011524781.2 | XP_011523083.1 | 5 | 1708 | |||
XM_017024609.1 | XP_016880098.1 | 7 | 1980 | |||
XM_017024610.1 | XP_016880099.1 | 6 | 1715 | |||
XM_017024611.1 | XP_016880100.1 | 6 | 1789 | |||
XM_017024612.1 | XP_016880101.1 | 5 | 1708 | |||
XM_017024613.1 | XP_016880102.1 | 6 | 1836 | |||
GenBank mRNA | AF153815.1 | - | 5 | 1609 | ||
AF153816.1 | - | 5 | 1609 | |||
AF153817.1 | - | 3 | 1381 | |||
AF179353.1 | - | 1 | 1123 | |||
AK225944.1 | - | 8 | 1634 | |||
AK290799.1 | - | 4 | 1478 | |||
BC033038.2 | - | 4 | 1615 | 5262150 |