Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ENPP2 SIRNA Assays ›
          Gene Symbol:
          ENPP2
          Gene Name:
          ectonucleotide pyrophosphatase/phosphodiesterase 2
          Gene Aliases:
          ATX, ATX-X, AUTOTAXIN, LysoPLD, NPP2, PD-IALPHA, PDNP2
          Chromosome Location:
          Chr.8: 119557077 - 119673404 on Build GRCh38
          UniGene ID:
          Hs.190977
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104805

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ENPP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040092.2 NP_001035181.1 8, 9 863
          NM_001130863.2 NP_001124335.1 8, 9 863
          NM_006209.4 NP_006200.3 8, 9 863
          XM_006716584.1 XP_006716647.1 8, 9 841
          XM_006716585.1 XP_006716648.1 8, 9 841
          XM_006716587.1 XP_006716650.1 8, 9 841
          XM_017013571.1 XP_016869060.1 8, 9 897
          XM_017013572.1 XP_016869061.1 8, 9 841
          XM_017013573.1 XP_016869062.1 8, 9 897
          XM_017013574.1 XP_016869063.1 8, 9 897
          XM_017013575.1 XP_016869064.1 8, 9 841
          GenBank mRNA AK315253.1 - 8, 9 836
          BC034961.2 - 8, 9 836 4828484
          D45421.1 - 8, 9 810
          EU131011.2 - 8, 9 777
          L35594.1 - 8, 9 826

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          phosphate-containing compound metabolic process
          receptor-mediated endocytosis
          movement of cell or subcellular component
          chemotaxis
          immune response
          G-protein coupled receptor signaling pathway
          phospholipid catabolic process
          positive regulation of epithelial cell migration
          regulation of cell migration
          phosphatidylcholine catabolic process
          regulation of angiogenesis
          positive regulation of peptidyl-tyrosine phosphorylation
          nucleic acid phosphodiester bond hydrolysis
          positive regulation of lamellipodium morphogenesis
          nucleic acid binding
          phosphodiesterase I activity
          nucleotide diphosphatase activity
          lysophospholipase activity
          scavenger receptor activity
          calcium ion binding
          transcription factor binding
          zinc ion binding
          hydrolase activity
          polysaccharide binding
          alkylglycerophosphoethanolamine phosphodiesterase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-996lf:80/100.66.79.239:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0