Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270476.1 | NP_001257405.1 | 16 | 2593 | ||
NM_006245.3 | NP_006236.1 | 16 | 2576 | |||
NM_180976.2 | NP_851307.1 | 16 | 2480 | |||
NM_180977.2 | NP_851308.1 | 14 | 2258 | |||
GenBank mRNA | AB209861.1 | - | 13 | 2101 | ||
AK122701.1 | - | 16 | 2470 | |||
BC001095.2 | - | 16 | 2460 | 3507280 | ||
BC001175.1 | - | 16 | 2387 | 3355395 | ||
BC010692.1 | - | 16 | 2470 | 3853892 | ||
D78360.1 | - | 16 | 2378 | |||
L76702.1 | - | 16 | 2584 |