Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KCNAB1 SIRNA Assays ›
          Gene Symbol:
          KCNAB1
          Gene Name:
          potassium voltage-gated channel subfamily A member regulatory beta subunit 1
          Gene Aliases:
          AKR6A3, KCNA1B, KV-BETA-1, Kvb1.3, hKvBeta3, hKvb3
          Chromosome Location:
          Chr.3: 156118211 - 156540603 on Build GRCh38
          UniGene ID:
          Hs.654519
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104877

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KCNAB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001308217.1 NP_001295146.1 13 2006
          NM_001308222.1 NP_001295151.1 13 2759
          NM_003471.3 NP_003462.2 14 2213
          NM_172159.3 NP_751891.1 14 2896
          NM_172160.2 NP_751892.1 14 2093
          XM_011513115.2 XP_011511417.1 14 1806
          XM_011513116.2 XP_011511418.1 12 1885
          XM_011513117.2 XP_011511419.1 12 1696
          XM_017007171.1 XP_016862660.1 14 1883
          XM_017007172.1 XP_016862661.1 14 3099
          XM_017007173.1 XP_016862662.1 12 1800
          GenBank mRNA AK057059.1 - 14 2631
          AK127240.1 - 14 2825
          BC043166.2 - 14 2843 5286833
          L39833.1 - 14 2086
          L47665.1 - 14 2080
          U33428.1 - 14 2004

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          potassium ion transport
          brain development
          heart development
          skeletal muscle tissue development
          learning or memory
          myoblast differentiation
          oxidation-reduction process
          diaphragm development
          potassium ion transmembrane transport
          regulation of potassium ion transmembrane transport
          regulation of delayed rectifier potassium channel activity
          negative regulation of delayed rectifier potassium channel activity
          aldo-keto reductase (NADP) activity
          voltage-gated potassium channel activity
          potassium channel regulator activity
          ion channel binding
          NADPH binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0