Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178019.2 | NP_821138.1 | 8 | 1338 | ||
GenBank mRNA | AF432876.1 | - | 8 | 1320 | ||
AK292427.1 | - | 8 | 1337 | |||
AY156951.1 | - | 4 | 540 | |||
BC110384.1 | - | 8 | 1338 | 5166215 |