Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN22 SIRNA Assays ›
          Gene Symbol:
          PTPN22
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 22
          Gene Aliases:
          LYP, LYP1, LYP2, PEP, PTPN8
          Chromosome Location:
          Chr.1: 113813811 - 113871761 on Build GRCh38
          UniGene ID:
          Hs.535276
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104901

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPN22
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193431.2 NP_001180360.1 2 242
          NM_001308297.1 NP_001295226.1 2 242
          NM_012411.5 NP_036543.4 2 242
          NM_015967.6 NP_057051.3 2 242
          XM_011541221.1 XP_011539523.1 2 223
          XM_011541222.1 XP_011539524.1 2 223
          XM_011541223.2 XP_011539525.1 2 223
          XM_011541225.2 XP_011539527.1 2 244
          XM_017001004.1 XP_016856493.1 2 223
          XM_017001005.1 XP_016856494.1 2 204
          XM_017001006.1 XP_016856495.1 2 223
          GenBank mRNA AF001846.1 - 2 147
          AF077031.1 - 2 178
          AF150732.1 - 2 176
          AK303124.1 - 2 191
          AK310570.1 - 2 195
          AK310698.1 - 2 195
          BC017785.1 - 2 195
          BC071670.1 - 2 181 5497108
          CD692658.1 - 2 262
          DB145424.1 - 3 421
          DN997966.1 - 2 152
          GU479452.1 - 2 156
          JN084012.1 - 2 119

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106460, XM_006233072, XM_008761379.

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          autophagy
          negative regulation of autophagy
          positive regulation of gene expression
          negative regulation of gene expression
          T cell differentiation
          lipopolysaccharide-mediated signaling pathway
          positive regulation of type I interferon production
          response to lipopolysaccharide
          negative regulation of tumor necrosis factor production
          regulation of natural killer cell proliferation
          positive regulation of toll-like receptor 3 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          peptidyl-tyrosine dephosphorylation
          phosphoanandamide dephosphorylation
          negative regulation of JUN kinase activity
          regulation of innate immune response
          regulation of B cell receptor signaling pathway
          negative regulation of T cell receptor signaling pathway
          negative regulation of T cell activation
          positive regulation of ERK1 and ERK2 cascade
          negative regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
          cellular response to muramyl dipeptide
          negative regulation of interleukin-6 secretion
          regulation of NIK/NF-kappaB signaling
          positive regulation of protein K63-linked ubiquitination
          positive regulation of interferon-gamma secretion
          negative regulation of p38MAPK cascade
          negative regulation of interleukin-8 secretion
          protein tyrosine phosphatase activity
          protein binding
          phosphatase activity
          SH3 domain binding
          kinase binding
          ubiquitin protein ligase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0