Search
Search

CD9| Gene Symbol: CD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001769.3 | NP_001760.1 | 8 | 1165 | ||
| XM_011521040.1 | XP_011519342.1 | 8 | 1144 | |||
| GenBank mRNA | AB079244.1 | - | 8 | 1084 | ||
| AY966455.1 | - | 8 | 1093 | |||
| BC011988.1 | - | 8 | 1032 | 3860667 | ||
| M38690.1 | - | 8 | 1030 | |||
| X60111.1 | - | 8 | 1092 | |||