Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INPP5K SIRNA Assays ›
          Gene Symbol:
          INPP5K
          Gene Name:
          inositol polyphosphate-5-phosphatase K
          Gene Aliases:
          PPS, SKIP
          Chromosome Location:
          Chr.17: 1494575 - 1516888 on Build GRCh38
          UniGene ID:
          Hs.632238
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104920

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: INPP5K
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135642.1 NP_001129114.1 5, 6 1048
          NM_016532.3 NP_057616.2 3, 4 658
          NM_130766.2 NP_570122.1 4, 5 889
          XM_005256683.2 XP_005256740.1 3, 4 498
          XM_011523934.1 XP_011522236.1 5, 6 671
          XM_011523936.2 XP_011522238.1 3, 4 498
          XM_017024755.1 XP_016880244.1 4, 5 729
          XM_017024756.1 XP_016880245.1 5, 6 806
          XM_017024758.1 XP_016880247.1 3, 4 428
          GenBank mRNA AB036829.1 - 3, 4 443
          AB036830.1 - 4, 5 672
          AK303907.1 - 3, 4 388
          AK312585.1 - 3, 4 443
          AK312844.1 - 3, 4 658
          BC004362.1 - 3, 4 457 3354658

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          negative regulation of protein phosphorylation
          regulation of glycogen biosynthetic process
          negative regulation of protein kinase activity
          phosphatidylinositol biosynthetic process
          G-protein coupled receptor signaling pathway
          negative regulation of peptidyl-threonine phosphorylation
          negative regulation of glucose transport
          dephosphorylation
          actin cytoskeleton organization
          cellular response to insulin stimulus
          cellular response to hormone stimulus
          negative regulation of peptidyl-serine phosphorylation
          negative regulation of dephosphorylation
          positive regulation of urine volume
          glucose homeostasis
          negative regulation of MAP kinase activity
          negative regulation by host of viral transcription
          negative regulation of glycogen biosynthetic process
          negative regulation of single stranded viral RNA replication via double stranded DNA intermediate
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          negative regulation of insulin receptor signaling pathway
          inositol phosphate dephosphorylation
          phosphatidylinositol dephosphorylation
          negative regulation of stress fiber assembly
          negative regulation of protein kinase B signaling
          negative regulation of calcium ion transport
          cellular response to cAMP
          cellular response to tumor necrosis factor
          cellular response to epidermal growth factor stimulus
          protein targeting to plasma membrane
          negative regulation of protein targeting to membrane
          ruffle assembly
          negative regulation of glycogen (starch) synthase activity
          positive regulation of renal water transport
          phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity
          inositol-polyphosphate 5-phosphatase activity
          vasopressin receptor activity
          protein binding
          inositol bisphosphate phosphatase activity
          phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
          phosphatidylinositol trisphosphate phosphatase activity
          phosphatidylinositol phosphate 5-phosphatase activity
          lipid phosphatase activity
          inositol trisphosphate phosphatase activity
          inositol-1,4,5-trisphosphate 5-phosphatase activity
          inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0