Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002800.4 | NP_002791.1 | 4, 5 | 462 | ||
GenBank mRNA | AK303118.1 | - | 3, 4 | 333 | ||
BC065513.1 | - | 4, 5 | 417 | 5749985 | ||
BI832878.1 | - | 4, 5 | 444 | |||
BX641100.1 | - | 2, 3 | 2143 | |||
CR541656.1 | - | 4, 5 | 393 | |||
S75169.1 | - | 3, 4 | 372 | |||
U01025.1 | - | 3, 4 | 372 | |||
X62741.1 | - | 4, 5 | 408 |