Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007058.3 | NP_008989.2 | 17, 18 | 1836 | ||
XM_006714984.1 | XP_006715047.1 | 17, 18 | 1839 | |||
XM_006714985.1 | XP_006715048.1 | 17, 18 | 1839 | |||
XM_006714987.1 | XP_006715050.1 | 17, 18 | 1839 | |||
XM_011514274.1 | XP_011512576.1 | 17, 18 | 1920 | |||
XM_011514275.1 | XP_011512577.1 | 18, 19 | 1962 | |||
GenBank mRNA | AJ242832.1 | - | 16, 17 | 1790 | ||
AK300107.1 | - | 8, 9 | 762 | |||
AK314054.1 | - | 17, 18 | 1816 | |||
BC033733.1 | - | 17, 18 | 1807 | 5170266 |