Search
Search

USP46| Gene Symbol: USP46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134223.1 | NP_001127695.1 | 9 | 2304 | ||
| NM_001286767.1 | NP_001273696.1 | 9 | 2192 | |||
| NM_001286768.1 | NP_001273697.1 | 10 | 2395 | |||
| NM_022832.3 | NP_073743.2 | 9 | 2228 | |||
| XM_017008557.1 | XP_016864046.1 | 9 | 2253 | |||
| GenBank mRNA | AK022614.1 | - | 9 | 2048 | ||
| AK024318.1 | - | 9 | 2169 | |||
| BC037574.1 | - | 9 | 2177 | 5529667 | ||