Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002428.3 | NP_002419.1 | 9, 10 | 2363 | ||
GenBank mRNA | BC006412.1 | - | 9, 10 | 1483 | ||
BC036495.1 | - | 9, 10 | 2369 | 5263145 | ||
BC055428.1 | - | 9, 10 | 2364 | 4811572 | ||
BT006715.1 | - | 7, 8 | 1263 | |||
D85510.1 | - | 6, 7 | 1093 | |||
D86331.1 | - | 8, 9 | 1301 | |||
Z48482.1 | - | 9, 10 | 1626 |